Would you like to log in again?
PGF2α High Sensitivity ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-931-069 ADI-930-069
89141-764EA 499 USD InStock
89141-764 89141-768
PGF2α High Sensitivity ELISA Kit, Enzo Life Sciences
Assays ELISAs
Most sensitive PGF2α ELISA kit available for prostaglandin research.
For the quantitative determination of PGF2α in culture supernatants, milk, plasma, serum, saliva, and urine from any species.
The PGF2α high sensitivity EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 3 hours. Absorbance is read at 405 nm. Simply the most sensitive quantification fo PGF2α commercially available in an enzyme immunoassay. Assay more samples without extraction.
Ordering information: Contains DxS IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty