Would you like to log in again?
PDI ELISA Development Set, Enzo Life Sciences
Supplier: Enzo Life Sciences
ImmunoSet®
ADI-960-072
89141-782EA 756 USD InStock
89141-782
PDI ELISA Development Set, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of PDI in cell lysates, culture supernatants, plasma, and tissue of human, mouse, and rat origin.
The ImmunoSet® PDI ELISA development set contains sufficient reagents for the development of 5 x 96 well ELISA plates which provides analysis of ~200 samples in duplicate. Save time, money, and sample by developing your own ELISA kits. Simply coat your plates and go! This kit provides superior accuracy with fully quantitative results compared to Western blot analysis and has low reactivity with related molecules.
Ordering information: Contains Capture antibody, Standard, Detection antibody, SA-HRP.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty