Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BAX Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-AAS-040-D ADI-AAS-040-F
89141-814EA 217 USD InStock
89141-814 89141-816
Anti-BAX Rabbit Polyclonal Antibody
Antibodies
Bax is an appromimately 22 kDa pro-apoptotic protein belonging to the Bcl-2 family of proteins, which are characterized by the presence of at least one Bcl-2 homology (BH) domains. Bax normally resides in the cytoplasm, but following an apoptotic stimuli, Bax undergoes a conformational change and translocates to mitochondrial membranes where it inserts and mediates the release of cytochrome c. This activates caspase-3 and perpetuates the apoptotic cascade.

  • For Western Blot

Recognizes human, mouse, rat, bovine, monkey, pig and rabbit Bax. Detects a band of ~22kDa by Western blot.

Type: Primary
Antigen: BAX
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty