Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EGFR Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ADI-CSA-330-E
89141-822EA 406 USD InStock
89141-822
Anti-EGFR Mouse Monoclonal Antibody
Antibodies
The epidermal growth factor receptor (EGFR) is a 170-kDa membrane bound receptor consisting of an extracellular ligand-binding domain, a single hydrophobic transmembrane region, and a tyrosine kinase domain-containing cytoplasmic region. Ligand binding induces receptor homo- or heterodimerization, triggering autophosphorylation of cytoplasmic tyrosine residues that provide docking sites for Src homology 2 (SH2) domain-containing signaling molecules. Src-mediated phosphorylation of EGFR at Tyr845 may contribute to EGFR hyperactivity in cancer, while autophosphorylation of EGFR at Tyr1068 and/or Tyr1173 in response to ligand binding is known to recruit cofactors (e.g. SHP1, RasGAP, SOS1, Grb2) that activate pathways associated with cell growth, apoptosis, adhesion, receptor endocytosis, and protein degradation.

  • For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes human EGF Receptor. Detects a band of ~170kDa by Western blot.

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty