Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFR1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-CSA-815-D ADI-CSA-815-F
89141-824EA 294 USD InStock
89141-824 89141-826
Anti-TNFR1 Rabbit Polyclonal Antibody
Antibodies
Tumor necrosis factor alpha (TNF-α), also known as cachectin, is a 17.5 kDa 157 amino acid member of the TNF superfamily of cytokines that is a potent lymphoid factor with effects on a wide range of target cells. Active TNFalpha is produced in both soluble and membrane-anchored trimers by macrophages, NK cells, and T- and B-lymphocytes. TNF exerts proinflammatory signals via binding and inducing trimerization of TNF-receptor1 (TNF-R1) expressed on most normal and transformed cells, or to TNF-receptor 2 (TNF-R2), expressed on endothelial and most immune cells. TNF signaling regulates hematopeiesis, differentation, endothelial cell activation, apoptosis, lipid metabolism, tumor progression, and immune suveillance, and dysregulation of TNF or its receptors is implicated in numerous disease states including cancer, osteoporosis, autoimmune disease, diabetes, and atherosclerosis.

  • For Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, bovine, dog, Drosophila, hamster, monkey, pig, rabbit, sheep, Xenopus and yeast TNF-R1. Detects a band of 55kDa by Western blot.

Type: Primary
Antigen: TNFR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty