Would you like to log in again?
DNA Damage ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-EKS-350
89141-832EA 1050 USD InStock
89141-832
DNA Damage ELISA Kit, Enzo Life Sciences
Assays ELISAs
Rapid DNA damage ELISA kit for cancer, apoptosis and oxidative stress.
For the measurement of DNA Damage (8-OHdG) in serum, saliva, and urine from any species. Cited sample types include plasma, seminal fluids, cell lysates, culture supernatants and DNA extracts.
The DNA Damage EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 2.5 hours. This kit has been validated in-house in a variety of sample matrices and is supplied in a convenient 96 well plate format. Protocol (xls) available upon request.
Ordering information: Contains Microtiter plate, Standard, Sample diluent, Wash buffer concentrate, Antibody, Antibody diluent, Conjugate, Conjugate diluent, TMB Substrate, Stop solution 2.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty