Would you like to log in again?
Anti-HSP60 IgG/A/M (human), ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-EKS-650
89141-856EA 962 USD InStock
89141-856
Anti-HSP60 IgG/A/M (human), ELISA Kit, Enzo Life Sciences
Assays ELISAs
Most comprehensive portfolio of heat shock protein (chaperone) products commercially available.
For the quantitative determination of anti-HSP60 IgG/A/M autoantibodies in human serum.
This assay allows for reproducible, accurate and precise determination of IgG , IgA and IgM antibodies (total) to human Hsp60 in serum. The ELISA kit uses recombinant human Hsp60 bound to the wells of the plate to bind anti-human Hsp60 antibodies present in human serum. The captured anti-human Hsp60 antibodies are detected with a HRP conjugated goat polyclonal antibody specific for human IgG, IgA and IgM molecules. The assay is developed with tetramethylbenzidine substrate pro ducing a blue color in proportion to the amount of captured anti -human Hsp60 antibodies. The color development is stopped with acid stop solution which converts the endpoint color to yellow. The intensity of the color is measured in a microplate reader at 450 nm.
Ordering information: Contains Microtiter plate, Standard, Sample diluent 1, Wash buffer concentrate, HRP Conjugate, TMB Substrate, Stop solution 2.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty