Would you like to log in again?
HSP70 ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-EKS-700B
89141-858EA 782 USD InStock
89141-858
HSP70 ELISA Kit, Enzo Life Sciences
Assays ELISAs
Widely used ELISA HSP70 kit for chaperone and stress research.
For the quantitative determination of HSP70 in cell lysates and tissue of human, mouse, and rat origin. Prod. No. ADI-EKS-715 is recommended for the detection of Hsp70 in serum and plasma samples.
The HSP70 EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4.5 hours. Absorbance is read at 450 nm. This kit is ideal for measuring intracellular HSP70 (HSP72). Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis.
Ordering information: Contains Microtiter plate, 5X Extraction reagent, Standard, Sample diluent 2, Wash buffer concentrate, Antibody, Conjugate, TMB Substrate, Stop solution 2.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty