Would you like to log in again?
HO-1 (human), ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-EKS-800
89141-866EA 1132.93 USD InStock
89141-866
HO-1 (human), ELISA Kit, Enzo Life Sciences
Assays ELISAs
Highly sensitive HO-1 ELISA kit enabling detection of HO-1 in cell lysates and tissue extracts in < 2.5 hours.
For the quantitative determination of human heme oxygenase-1 in cell lysates and tissue extracts. Cited sample types include serum and plasma.
The HO-1 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 2.5 hours. Absorbance is read at 450 nm. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis.
Ordering information: Contains Microtiter plate, 5X Extraction reagent, Standard, Sample diluent, Wash buffer concentrate, Antibody, Antibody diluent, HRP Conjugate, Conjugate diluent, TMB Substrate, Stop solution 2.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty