Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CAMK2 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
KAM-CA002-D KAM-CA002-F
89141-904EA 265 USD InStock
89141-904 89141-906
Anti-CAMK2 Mouse Monoclonal Antibody
Antibodies
CaMKIIa, the alpha subunit of Ca2+ calmodulin-dependent protein kinase II is part of a family of multifunctional protein kinases, which play a major role in Ca2+-mediated signal transduction.  CaMKIIa is expressed in many different tissues but is specifically found in the neurons of the forebrain and its mRNA is found within the dendrites as well as the soma of the neuron.  The neuronal CaMKII consists of two major subunits of 52 and 60 kDa which are encoded by a- and b-CaMKII genes, respectively.  Additional isoforms are generated by alternative splicing of these as well as of the ubiquitious g- and d-CaMKII genes.  Each subunit has an ATP-binding domain (arginine-X-X-serune/threonine), consensus phosphorylation site, catalytic domain, and a centrally located regulatory domain which has calmodulin binding activity. Activation and autophosphorylation of CaMKII may regulate numerous neuronal processes which includes two forms of synaptic plasticity, long term potentiation and long term depression.  Neuronal CaMKII subunits assemble as large multimeric holoenzymes.  The C-terminal association domains of  6-12 subunits assemble into a central globular structure from which the N-terminal catalytic/regulatory domains extend radially like petals of a flower. The subunit composition of the rat forebrain CaMKII holoenzyme consists of heteromers composed of a and β subunits at a ratio of 2:1 and homomers composed of only a subunits.  The association of CaMKII subunits leads to the positioning of their catalytic/regulatory domains in close proximity and the neighboring calmodulin-bound subunits cooperate to rapidly phosphorylate each other.  Autophosphorylation also enables CaMKII to attain an enhanced affinity for NMDA receptors in postsynaptic densities.

  • For Immunohistochemistry, Western Blot, in vitro Assay

Recognizes mouse and rat CaMKII. Detects a band of ~50-60kDa by Western blot.

Type: Primary
Antigen: CAMK2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty