Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RAP1A Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAP-GP126-D
89141-908EA 301 USD InStock
89141-908
Anti-RAP1A Rabbit Polyclonal Antibody
Antibodies
The Ras superfamily of small guanosine triphosphatases (GTPases) is traditionally subdivided into five families: Ras, Rho, Rab, Ran, and Arf. Rap1/Krev1 is a member of the Ras family of low molecular weight GTP-binding proteins1. Ras-like GTPases are ubiquitously expressed, evolutionarily conserved molecular switches that couple extracellular signals to various cellular responses. Rap1 is primarily found at the cytosolic side of intracellular membranes and has two isoforms: Rap1a and 1b2. Both isoforms have a molecular mass of 21 kDa and are isoprenylated at the carboxyl-terminal and phosphorylated by the cAMP-dependent protein kinase A (PKA)3. Rap1 cycles between a GTP-bound active form and a GDP-bound inactive form that is mediated by GTPase activating protein (GAP) and GDP dissociation stimulator (GDS)4,5. Activation occurs by a variety of extracellular stimuli through several conserved guanine nucleotide exchange factors (GEFs) and GTPase activating proteins (GAPs). Rap1 is proposed to regulate Ras mediated signaling and may also be involved in the regulation of integrinmediated cell adhesion, although the mechanism of regulation is not known. Overexpression of Rap reverses the transformed phenotype induced by Ras, possibly by competing with Ras for interaction with Ras-GAP. Rap has been shown to participate in MAP kinase cascade activated by growth factor and maintaining human T cell anergic state by blocking IL-2 expression.

  • For Western Blot

Recognizes human, mouse, rat, bovine, dog, guinea pig and rabbit Rap1. Detects a band of ~25kDa by Western blot.

Type: Primary
Antigen: RAP1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty