Would you like to log in again?
Liver (Human), Microsome Extract, Enzo Life Sciences
Supplier: Enzo Life Sciences
LYT-HM100-F
89141-960EA 148 USD InStock
89141-960
Liver (Human), Microsome Extract, Enzo Life Sciences
Specimens Preserved Specimens Preserved Animal Materials
Liver microsomes are subcellular particles derived from the endoplasmic reticulum of hepatic cells
Liquid. In SDS-PAGE sample buffer, pH 8.8, containing 40 mM TRIS-hydrogen chloride, 1% SDS, 50mM DTT, 7.5% glycerol, and 0.003% Bromophenol Blue and concentration is 2 mg/mL.
Microsomes are a rich source of heme-containing metabolic enzymes, including heme-oxygenases and cytochrome P450 (CYP) enzymes, and are commonly used in the study of xenobiotic drug metabolism. This microsome preparation consists of microsomes pooled from various human donors of mixed genders
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty