Would you like to log in again?
Brain (Rat), (Tissue Extract), Enzo Life Sciences
Supplier: Enzo Life Sciences
LYT-RB100-F
89141-966EA 246 USD InStock
89141-966
Brain (Rat), (Tissue Extract), Enzo Life Sciences
Specimens Preserved Specimens Preserved Animal Materials
Liquid.
In SDS-PAGE sample buffer, pH 6.8, containing 40 mM TRIS-HCl, 1% SDS, 50 mM DTT, 7.5% glycerol, and 0.003% bromophenol blue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty