Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CRYAB Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-227-F ADI-SPA-227-D
89142-046EA 639 USD InStock
89142-046 89142-044
Anti-CRYAB Rabbit Polyclonal Antibody
Antibodies
α-Crystallins composed of αA (~19 kDa) and αB (~19.2 kDa) subunits, are major water-soluble proteins accounting for almost 50% of total protein in the mammalian transparent eye lens and they are also found in a variety of other tissues. Alpha-crystallins are also referred to as small heat shock proteins, since they are induced by increased temperature in a variety of organisms. The α-crystallins have sequence homology as well as structural and functional similarities with the small Hsp’s such as Hsp25/27. Most α-crystallins have four common structural and functional features: (i) molecular weight between 12 and 43kDa; (ii) the formation of large oligomeric complexes composed of αA-crystallin, αB-crystallin and Hsp25/27; (iii) the moderately conserved α-crystallin domain in the central region of the protein; and (iv) molecular chaperone activity. The α-crystallin domain comprises approximately 90 residues, is bounded by variable N-terminal and C-terminal extensions and is involved in oligomer assembly. Oligomers can reach 800 kDa or more and are dynamic, exhibiting subunit exchanges and organizational plasticity, possibly leading to functional diversity. Phosphorylation of serine residues occurs during development and in response to stress, and usually decreases oligomer size. Chaperone activity requires, and is modulated by, oligomerization and is limited to binding unfolded intermediates to prevent irreversible aggregation. Although productive release and refolding of denatured proteins requires close cooperation with other chaperones. Other proposed functions include a role in membrane stabilization and modulation of intermediate filament organization during physiological stress and neurodegenerative disease. Alpha B Crystallin is phosphorylated on serine 45 by ERK1/2 and serine 59 by MAPKAPK-2. When mono-phosphoylated αB-crystallin is isolated it contains only αB-crystallin phosphorylated at serine 19.

  • For Immunoprecipitation, Western Blot

Recognizes mouse, rat, bovine, pig and sheep alphaB-crystallin.

Type: Primary
Antigen: CRYAB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty