Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HDJ1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-400-F ADI-SPA-400-D
89142-050EA 481 USD InStock
89142-050 89142-048
Anti-HDJ1 Rabbit Polyclonal Antibody
Antibodies
Hsp40/Hdj1 is a cytosolic co-chaperone belonging to the class DnaJ, named after its homolog in E. coli. Hsp40 functions in protein folding by binding nascent peptides and unfolded substrates and facilitating substrate interaction with Hsp70. Binding of Hsp40 to Hsp70 increases Hsp70 ATP hydrolysis and substrate binding, which is reversed by the action of nucleotide exchange factors (GrpE in E. coli). Repeated cycles of Hsp40/Hsp70 peptide binding and ATP hydrolysis prevent premature folding and aggregation, promoting protein maturation throughout the cell.

  • For Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, bovine, beluga, dog, chicken, fish, guinea pig, hamster, mouse, monkey, mussel, pig, rabbit, sheep, scallop and Xenopus HSP40 (Hdj1). Detects a band of ~40kDa by Western blot.

Type: Primary
Antigen: HDJ1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty