Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Glutathione Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-542-E
89142-052EA 442 USD InStock
89142-052
Anti-Glutathione Mouse Monoclonal Antibody
Antibodies
Glutathione, a major intracellular non-protein thiol, protects against free radical damage by providing reducing equivalents for several key antioxidant enzymes. In addition, glutathione acts as a scavenger of hydroxyl radicals and nascent oxygen. Elevated glutathione levels are associated with a reduced rate of illness, higher levels of self-rated health, lower cholesterol, lower body mass index, and lower blood pressure among the elderly. Glutathione provides a primary defense system for the removal of oxidants in the brain. Studies have revealed a correlation between low glutathione levels and damage to neurons that manufacture dopamine, suggesting a link to Parkinson's disease.

  • For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot

Species independent.

Type: Primary
Antigen: GLUTATHIONE
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Species Independent

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty