Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ERP57 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-585-D ADI-SPA-585-F
89142-056EA 500 USD InStock
89142-056 89142-054
Anti-ERP57 Rabbit Polyclonal Antibody
Antibodies
ERp57 (also known as Grp58), a member of the Thioredoxin superfamily, is an ER protein with disulfide isomerase activity. It has been shown to increase after oncogenic transformation and to be a component of the MHC Class I peptide-loading complex.

  • For Western Blot

Recognizes human, mouse, rat, bovine, dog, guinea pig, hamster, monkey, pig and rabbit ERp57. Detects a band of ~57kDa by Western blot.

Type: Primary
Antigen: ERP57
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty