Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSP70 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-8131-D ADI-SPA-8131-F
89142-064EA 531 USD InStock
89142-064 89142-062
Anti-HSP70 Mouse Monoclonal Antibody
Antibodies
The 70 kDa heat shock protein Hsp70 belongs to the Hsp70 family of highly-related protein isoforms ranging in size from 66 kDa to 78 kDa. Hsc70 shares close biochemical and biological ties to Hsp70, and also belongs to the Hsp70 family. These proteins include cognate members found within major intracellular compartments and highly inducible isoforms predominantly cytoplasmic or nuclear in distribution. Members of the Hsp70 family function as molecular chaperones involved in such cellular functions as protein folding, transport, maturation and degradation, operating in an ATP-dependent manner. The molecular chaperones of the Hsp70 family recognize and bind to nascent polypeptide chains or partially folded intermediates of proteins, preventing their aggregation and misfolding, and the binding of ATP triggers a critical conformational change leading to the release of the bound substrate protein. Data demonstrates that with a ubiquitin-like domain at its amino terminus and its association with the 26S proteosome in HeLa cells, Bag-1 modulates the chaperone activity of Hsc70 and Hsp70. These findings reveal Bag-1's role as a physical link between the Hsc70/Hsp70 chaperone system and the proteasome. Experimental data also shows that the ATPase domain and the substrate-binding domain of Hsp70 (or Hsc70) cooperate to form a co-chaperone-chaperone complex with the synaptic vesicle cysteine string protein (csp), essential for normal neurotransmitter release.

  • For Western Blot

Recognizes human and bacteria HSP70 (HSP72). Detects a band of ~70kDa by Western blot.

Type: Primary
Antigen: HSP70
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty