Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSF1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-902-F ADI-SPA-902-D
89142-100EA 639 USD InStock
89142-100 89142-098
Anti-HSF1 Rabbit Polyclonal Antibody
Antibodies
HSFs (Heat Shock family of transcription factors), which consists of HSF 1-4, bind to highly conserved Heat shock elements (HSEs) in the promoter regions of heat shock genes, ultimately regulating the expression of Heat shock proteins (Hsps). On exposure to heat shock and other stresses, HSF1 localizes within seconds to discrete nuclear granules and on recovery from stress, HSF1 rapidly dissipates from the stress granules to a diffuse nucleoplasmic distribution.

  • For Western Blot

Recognizes human HSF1. Predicted MW of ~60kDa. Detects a band of 85-95kDa by Western blot depending on hyperphosphorylation of the native protein.

Type: Primary
Antigen: HSF1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty