Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RSEC8 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
VAM-SV016-D VAM-SV016-F
89142-316EA 590 USD InStock
89142-316 89142-314
Anti-RSEC8 Mouse Monoclonal Antibody
Antibodies
rSec6 and rSec8 are rat homologs of the yeast Sec6 and Sec8 proteins that are essential for protein transport between the Golgi apparatus and the plasma membrane. In yeast, Sec8 is part of a 19.5S complex that includes Sec6 and Sec15, that also associate with the ras-like GTP binding protein Sec4. An analogous 17S complex in rat brain was found to contain rSec6 and rSec8, along with eight proteins with a combined molecular weight of 743 kDa. The membrane-associated rSec6/8 complex co-immunoprecipitates with syntaxin, providing evidence that this complex is involved in synaptic vesicle docking and fusion at the presynaptic terminal.

  • For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, bovine, dog, chicken, fish, guinea pig, hamster, monkey, pig, rabbit and sheep rSec8. Detects a band of ~110kDa by Western blot.

Type: Primary
Antigen: RSEC8
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty