Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-STX6 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
VAM-SV025-F VAM-SV025-D
89142-318EA 277 USD InStock
89142-318 89142-320
Anti-STX6 Mouse Monoclonal Antibody
Antibodies
Syntaxins are type II integral membrane proteins localized to membranes that regulate vesicle transport, protein maturation, and exocytosis. Syntaxin 1A resides in the nerve terminals of sensory neurons and nerve fibers reaching small blood vessels. Syntaxin 2 localizes to the apical plasma membrane and regulates epithelial-mesenchymal interactions. Syntaxin 6 is primarily localized to the trans-Golgi network, and partially co-localizes with AP-1 on clathrin-coated membranes. Syntaxin 13 is ubiquitously expressed and localizes to the tubular early and recycling endosomes, as well as endosomal vacuoles.

  • For Immunoprecipitation, Western Blot

Recognizes human, mouse, rat and hamster Syntaxin 6. Detects a band of ~35kDa by Western blot.

Type: Primary
Antigen: STX6
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty