Would you like to log in again?
Nampt (Visfatin/PBEF) (Mouse/Rat) Dual ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
45A-0007EKKI01
89142-354EA 1310 USD InStock
89142-354
Nampt (Visfatin/PBEF) (Mouse/Rat) Dual ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of Nampt (Visfatin/PBEF) in mouse or rat serum.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty