Would you like to log in again?
Autocamtide-2-Related Inhibitory Peptide
Supplier: Enzo Life Sciences
151-029-M005 151-029-M001
89143-030EA 501.86 USD InStock
89143-030 89143-028
Autocamtide-2-Related Inhibitory Peptide
Proteins and Peptides
CaM kinase II inhibitor
Highly specific and potent inhibitor of Ca2+/calmodulin-dependent protein kinase II without affecting cAMP-dependent protein kinase (PKA), protein kinase C (PKC) and Ca2+/calmodulin-dependent protein kinase IV.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty