Would you like to log in again?
Nociceptin
Supplier: Enzo Life Sciences
163-001-P005 163-001-P001
89143-610EA 472.93 USD InStock
89143-610 89143-608
Nociceptin
Proteins and Peptides
ORL1 receptor agonist
ORL1 receptor agonist. The ORL1 receptor structurally resembles opioid receptors and is negatively coupled with adenylyl cyclase.
Ordering information: Quantity: *Sold as net peptide weight
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty