Would you like to log in again?
Human Recombinant Caspase-6 (from E. coli)
Supplier: Enzo Life Sciences
201-060-U025 201-060-U100
89144-016EA 675 USD InStock
89144-016 89144-014
Human Recombinant Caspase-6 (from E. coli)
Enzymes
Produced in E. coli.
Cleaves substrates exhibiting consensus sequence VEID, e.g. Ac-VEID-AFC (Prod. No. ALX-260-111), Ac-VEID-AMC (Prod. No. ALX-260-064) and Ac-VEID-pNA (Prod. No. ALX-260-063).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty