Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ESR2 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-135-C050
89145-214EA 811 USD InStock
89145-214
Anti-ESR2 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunocytochemistry, Immunohistochemistry, Western Blot, Gel Shift Assay

Recognizes human, mouse, rat, sheep and monkey ERbeta. Detects a band of ~55kDa by Western blot. Does not cross-react with ERalpha.

Type: Primary
Antigen: ESR2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty