Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LEPR Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-168-R100
89145-270EA 810 USD InStock
89145-270
Anti-LEPR Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Western Blot

Recognizes human, mouse and rat leptin receptor. Detects bands of ~100kDa and ~125kDa by Western blot, representing the short and long forms of leptin receptor. Also detects two unrelated bands of ~31kDa and ~35kDa.

Type: Primary
Antigen: LEPR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty