Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Peroxisomal Membrane Protein 70 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-205-C100
89145-306EA 874 USD InStock
89145-306
Anti-Peroxisomal Membrane Protein 70 Rabbit Polyclonal Antibody
Antibodies
Peroxisomes are single membrane organelles which are involved in many important biochemical pathways and are found in almost all eukaryotic cells. Various toxins including phenols, alcohols, and aldehydes are targeted for modification by peroxisomes. Long chain fatty acid metabolism via β-oxidation is also mediated by this organelle. Loss of peroxisomal functions may result in diseases such as Zellweger syndrome, hyperpipecolic acidemia, neonatal adrenoleukodystrophy, and infantile Refsums disease. Peroxisomal membrane protein 70 (PMP70) is an abundant, integral membrane protein of the peroxisome. This protein is induced by treatment with hypolipidemic agents in parallel with peroxisome proliferation and stimulation of the peroxisomal β-oxidation enzymes.

  • For Western Blot

Recognizes mouse PMP70. Detects a band of ~70kDa by Western blot.

Type: Primary
Antigen: PEROXISOMAL MEMBRANE PROTEIN 70
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty