Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Bone Sialoprotein 2 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-312-R020 210-312-R100
89145-462EA 280 USD InStock
89145-462 89145-464
Anti-Bone Sialoprotein 2 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes human and rat bone sialoprotein. Does not cross-react with human osteopontin or osteonectin.

Reconstitute with 20μl or 100μl distilled water.

Type: Primary
Antigen: BONE SIALOPROTEIN 2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty