Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ETRA Sheep Polyclonal Antibody

Supplier: Enzo Life Sciences
210-507A-C250
89145-830EA 529 USD InStock
89145-830
Anti-ETRA Sheep Polyclonal Antibody
Antibodies
Endothelins are a family of peptides that are potent vasoconstrictors produced by mammalian vascular endothelial cells. The receptors for the endothelins are found on various tissues including, but not restricted to, lung and intestine.

  • For Immunohistochemistry, Western Blot

Recognizes an epitope on the cytoplasmic side of human, mouse and rat ETA-receptor. Detects bands of 34-52kDa. Does not block the binding of endothelins to the receptor.

Type: Primary
Antigen: ETRA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty