Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ENOS Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-509-R100
89145-834EA 813 USD InStock
89145-834
Anti-ENOS Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse, rat, bovine, pig and dog eNOS (NOS III). Detects a band of ~140kDa by Western blot. Does not cross-react with human iNOS (NOS II) or rat nNOS (NOS I).

Type: Primary
Antigen: ENOS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty