Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SHC1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-535-R100
89145-878EA 285 USD InStock
89145-878
Anti-SHC1 Rabbit Polyclonal Antibody
Antibodies
There are 3 alternatively spliced isoforms of the Shc adapter pro-tein: p46SHC, p52SHC and p66SHC, that link activated growth factor receptors to signalling pathway. Isoform p46SHC and p52SHC, once phosphorylated, couple activated receptor tyrosine kinases to Ras via the recruitment of the GRB2/SOS com-plex and are implicated in mitogenic signalling. They function as initiators of the Ras signalling cascade in various non-neuronal systems. The isoform p66SHC does not mediate Ras activation, but is involved in signal transduction pathways that regulate the cellular response to oxidative stress and life span. It acts as a downstream target of the tumor suppressor p53 and is indispen-sable for the ability of stress-activated p53 to induce elevation of intracellular oxidants, cytochrome c release and apoptosis.

  • For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot

Recognizes human, mouse and hamster p46shc, p52shc and p66shc isoforms of phosphorylated and unphosphorylated Shc.

Type: Primary
Antigen: SHC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty