Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ITGAV Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-537-R100
89145-882EA 433 USD InStock
89145-882
Anti-ITGAV Rabbit Polyclonal Antibody
Antibodies
CD51 (integrin αV subunit) associates with CD61 (integrin β3 subunit) binding to the RGD sequence in extracellular matrix proteins like vitronectin, fibrinogen, thrombospondin, von Willebrand factor and osteopontin. The αV/β3 heterodimer is expressed in endothelial cells, leukocytes, NK cells, macrophages, neutrophils, platelets and several malignant tumor cells and seems to play a role in embryo implantation, angiogenesis, wound healing and cancer. αV/β3  is also highly expressed in osteoclasts being important for bone resorption.

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes cytoplasmic domain of CD51 of all species.

Type: Primary
Antigen: ITGAV
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty