Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDH2 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-618-C400
89145-950EA 1050 USD InStock
89145-950
Anti-CDH2 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse, bovine and chicken N-cadherin. Does not cross-react with human E-or P-cadherin.

Reconstitute with 200µl distilled water. Dilute with 20mM TBS, pH 7.4, containing 1% BSA and 0.1% sodium azide and 10mM calcium chloride.  

Type: Primary
Antigen: CDH2
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty