Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ADIPOR2 Goat Polyclonal Antibody

Supplier: Enzo Life Sciences
210-647-C200
89146-008EA 547 USD InStock
89146-008
Anti-ADIPOR2 Goat Polyclonal Antibody
Antibodies
Host: Goat

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes mouse AdipoR2.

Type: Primary
Antigen: ADIPOR2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty