Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GUCY1A3 Chicken Polyclonal Antibody

Supplier: Enzo Life Sciences
210-786-R100
89146-064EA 465 USD InStock
89146-064
Anti-GUCY1A3 Chicken Polyclonal Antibody
Antibodies
Host: Chicken

  • For Western Blot

Recognizes endogenous rat sGCalpha1beta1. Recognizes human and mouse sGCbeta1. Does not cross-react with human and mouse sGCalpha1.

Type: Primary
Antigen: GUANYLYL CYCLASE ALPHA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty