Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-8-Hydroxyguanosine Goat Polyclonal Antibody

Supplier: Enzo Life Sciences
210-802-R100
89146-090EA 301 USD InStock
89146-090
Anti-8-Hydroxyguanosine Goat Polyclonal Antibody
Antibodies
8-Hydroxydeoxyguanosine (8OHdG) is a modified base that occurs in DNA due to attack by hydroxyl radicals that are formed as byproducts and intermediates of aerobic metabolism and during oxidative stress. There is increasing evidence to support the involvement of free radical reactions in the damage of biomolecules that eventually lead to several diseases in humans, such as atherosclerosis, cerebral and heart ischemia-reperfusion injury, cancer, rheumatoid arthritis, inflammation, diabetes, aging and neurodegenerative conditions, such as Alzheimer's disease. 8OHdG has become increasing popular as a sensitive, stable and integral marker of oxidative damage in cellular DNA. Biomonitoring in humans has demonstrated that 8OHdG can be excreted in the urine and that a significant increase is caused by exposure to tobacco smoke and ionizing radiation. Because 8OHdG is so well correlated with oxidative stress and damage to DNA, which leads to degenerative disease states, the development of an antibody that can be used to study DNA damage has numerous applications. In addition to the direct study of DNA damage within cells, this antibody has applications in the development of immunoassays that can monitor 8OHdG excretion in the urine and serve as a biomarker of oxidative stress. Industrial uses may extend to the dietary supplement manufacturers, who could benefit from an immunoassay that could be used to test the effectiveness of antioxidants and other nutraceuticals.

  • For ELISA, Flow Cytometry, Immunohistochemistry

Recognizes 8-hydroxyguanosine and 8-hydroxy-2'-deoxyguanosine. Partially cross-reacts with 8-mercaptoguanosine and slightly with 8-bromoguanosine. Does not cross-react with other naturally occuring nucleotides.

Type: Primary
Antigen: 8-HYDROXYGUANOSINE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Species Independent

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty