Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VASP Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-880-C025
89146-214EA 621 USD InStock
89146-214
Anti-VASP Rabbit Polyclonal Antibody
Antibodies
VASP (vasodilator stimulated phosphoprotein) is a proline-rich protein substrate of cAMP- and cGMP-dependent protein kinases. Phosphorylation of VASP at Ser-157 causes a mobility shift in SDS gel electrophoresis from 46 to 50 kDa, which has been used as a convenient marker to monitor cyclic nucleotide-dependent protein kinase activity. VASP is the founding member of the Ena-VASP protein family, comprising the Drosophila protein Enabled (Ena), its mouse homologue Mena (mammalian Enabled), and mouse EVL (Ena-VASP-like protein). With these proteins VASP shares a conserved overall domain organization:
a) the conserved N-terminal Ena-VASP homology domain 1 (EVH1), which mediates binding to a proline-rich motif
b) a more divergent proline-rich central domain (which is responsible for profilin binding)
c) a conserved C-terminal EVH2 domain.

VASP is expressed in a variety of mammalian cell types and tissues. In cultured cells, VASP is associated with focal adhesions, cell-cell contacts, microfilaments, and highly dynamic membrane regions. From in vitro binding data VASP has been suggested to link profilin to zyxin, vinculin, and the Listeria spp. surface protein ActA, respectively. Functional evidence indicates that VASP is a crucial factor involved in the enhancement of actin filament formation and the actin-dependent motility of intracellular bacterial pathogens.

  • For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse and pig VASP. Detects both the 46kDa (Ser157 dephospho) and 50kDa (Ser157 phospho) forms of VASP.

Type: Primary
Antigen: VASP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty