Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CRADD Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-915-C050
89146-276EA 309 USD InStock
89146-276
Anti-CRADD Rabbit Polyclonal Antibody
Antibodies
RAIDD (RIP-associated ICH-1 homologous protein with a death domain) is an adaptor molecule that mediates the action of cysteine proteases involved in apoptosis. In human cell lines, the caspase-2 adaptor RAIDD interacts selectively with caspase-2 through its caspase recruitment domain (CARD) and leads to caspase-2-dependent death. RAIDD overexpression induced caspase-2, CARD- and caspase-9-dependent apoptosis of PC12 cells and sympathetic neurons. Apoptosis correlated with the formation of discrete perinuclear aggregates. Both death and aggregates required the expression of full-length RAIDD. It has been reported that activation of caspase-2 occurs in a complex that contains the death domain–containing protein PIDD, whose expression is induced by p53, and the adaptor protein RAIDD.

  • For Immunoprecipitation, Western Blot

Recognizes human RAIDD.

Type: Primary
Antigen: CRADD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty