Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD39L1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
215-045-R100
89146-508EA 385 USD InStock
89146-508
Anti-CD39L1 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunocytochemistry

Recognizes human CD39L1.

Type: Primary
Antigen: CD39L1
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty