Would you like to log in again?
Dnp-Pro-Leu-Gly-Met-Trp-Ser-Arg-OH
Supplier: Enzo Life Sciences
260-120-M005
89146-892EA 334.08 USD InStock
89146-892
Dnp-Pro-Leu-Gly-Met-Trp-Ser-Arg-OH
Proteins and Peptides
Substrate for MMP-2 and -9
Fluorogenic substrate for MMP-2 and MMP-9. Excitation: 280nm; emission 360nm.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty