Would you like to log in again?
Ebelactone A ≥99%
Supplier: Enzo Life Sciences
270-013-M001
89147-046EA 136.66 USD InStock
89147-046
Ebelactone A ≥99%
Ebelactone A
Esterase inhibitor
Inhibitor of esterases, lipases and N-formylmethionine aminopeptidase.
Microbial.
CAS Number: 76808-16-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/55/45347455.jpg
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty