Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

NU 2058

Supplier: Enzo Life Sciences
Synonyms: O-Cyclohexylmethylguanine

270-394-M025 270-394-M005
89147-794EA 207.91 USD InStock
89147-794 89147-796
NU 2058
NU 2058
Competitive inhibitor of CDK1/cyclin B (IC50=5µM) and CDK2/cyclin A (IC50=12µM). Pharmacologically distinct from the relative specific inhibitor of CDK1/2 roscovitine.

  • CDK inhibitor

Pharmacologically distinct from the relative specific inhibitor of CDK1/2 roscovitine (Prod. No. BML-CC205).

Soluble in water or DMSO.


Formula: C₁₂H₁₇N₅O
MW: 247.30 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05664734
CAS Number: 161058-83-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/75/50/45347550.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty