Would you like to log in again?
Microcystis aeruginosa Microcystin LF
Supplier: Enzo Life Sciences
350-081-C025 350-081-M001 350-081-C100
76417-954EA 4529.24 USD InStock
76417-954 89149-022 89149-024
Microcystis aeruginosa Microcystin LF
Proteins and Peptides
Microcystins are a group of cyclic heptapeptide hepatotoxins produced by a number of cyanobacterial genera. The most notable of which, and namesake, is the widespread genus Microcystis. Structurally, all microcystins consist of the generalized structure cyclo(-D-Ala1-X2-D-MeAsp3-Y4-Adda5-D-Glu6-Mdha7-). X and Y are variable L-amino acids, D-MeAsp is D-erythro-ß-methylaspartic acid and Mdha is N-methyldehydroalanine. Adda is the cyanobacteria unique C20 ß-amino acid 3-amino-9-methoxy-2,6,8-trimethyl-10-phenyl-deca-4,6-dienoic acid. Substitutions of the variable L-amino acids at positions 2 and 4 give rise to at least 21 known primary microcystin analogs and alterations in the other constituent amino acids result in more than 90 reported mycrocystins to date.
Analog of microcystin-LR (Prod. No. ALX-350-012) with Phe substituted in place of Arg. Hydrophobic and believed to be more cell permeable than other microcystins. More toxic than the more hydrophilic analogs such as microcystin-LR and microcystin-LY.
Isolated from Microcystis aeruginosa.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty