Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lupeol ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Synonyms: Monogynol BFagarasterol20(29)-Lupen-3β-ol, 3β-Hydroxy-20(29)-lupeneClerodol

350-355-M050 350-355-M010 350-355-G001
89149-572EA 1637.94 USD InStock
89149-572 89149-574 89149-576
Lupeol ≥98% (by HPLC)
Lupeol
Anti-inflammatory, antioxidant and antimutagenic compound

Anti-inflammatory, antioxidant and antimutagenic compound. Shows preventive effects on DMBA induced DNA alkylation damage in mouse skin. Modulator of NF-κB and PI(3)K/Akt (protein kinase B; PKB) pathways. Induces Fas-mediated apoptosis via inhibition of Ras signalling. Inhibits tumor growth in mouse in vivo.

Isolated from Lupinus polyphyllus.


Formula: C₃₀H₅₀O
MW: 426.7
CAS Number: 545-47-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/77/58/45347758.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty