Synonyms:
Dodecyl-β-D-maltopyranoside
, (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
, N-Dodecyl-Beta-Maltoside
, Lauryl-β-D-maltoside
, DDM
500-007-G005
500-007-G025
500-007-G001
89151-634EA
175.95
USD
InStock
89151-634
89151-636
89151-638
n-Dodecyl-β-D-maltoside ≥99% (by HPLC), high purity
n-Dodecyl-β-D-maltoside
Non-ionic detergent free of alpha-isomer (HPLC)
- Formulation: White crystal powder.
Non-ionic detergent similar to n-octyl-beta-D-glucopyranoside (Prod. No. ALX-500-001). However, increased alkyl chain length has been shown to increase thermal stability of lipid-free rhodopsin in solution. Applicable to membrane-protein solubilization studies.