Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Recombinant MegaCD40L™ (soluble) (from CHO Cells)

Supplier: Enzo Life Sciences
522-110-C010
89151-892EA 989.5 USD InStock
89151-892
Human Recombinant MegaCD40L™ (soluble) (from CHO Cells)
Proteins and Peptides
High-activity, high-purity CD40L construct for immune activation. Produced in CHO cells. The extracellular domain of human CD40L (CD154) (aa 116-261) is fused at the N-terminus to mouse ACRP30headless (aa 18-111) and a FLAG®-tag.

MegaCD40L™ is a high-activity construct in which two trimeric CD40 ligand molecules are artificially linked via the collagen domain of Adiponectin/ACRP30/AdipoQ. This construct very effectively simulates the natural membrane-assisted aggregation of CD40L in vivo. It provides a simple and equally potent alternative to [CD40L+enhancer] combinations (Prod. No. ALX-850-064).

Produced in CHO cells. The extracellular domain of human CD40L (CD154) (aa 116-261) is fused at the N-terminus to mouse ACRP30headless (aa 18-111) and a FLAG®-tag.

* Purity - 10- to 100-fold lower Endotoxin levels than comparable products
* Activity – MegaCD40L™ oligomer mimics in vivo membrane-assisted CD40L aggregation and stimulation without the need for enhancers
* Consistency – Experienced manufacturing results in dependable lot-to-lot performance
* Stability – Optimized formulation for improved stability and enhanced activity

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty