Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

APF ~5 mM ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Danger
Synonyms: Aminophenyl fluorescein

620-075-M001
89152-914EA 709.04 USD InStock
89152-914
APF ~5 mM ≥98% (by HPLC)
APF
Fluorescent probe for hROS detection.

Fluorescent reagent (Ex(max): 490 nm; Em(max): 515 nm) for the detection of highly reactive oxygen species (hROS). Immediately reacts with hROS such as hydroxyl radical, peroxynitrite and hypochlorite, and the fluorescence intensity greatly increases. Use of APF together with HPF also allows for specific detection of hypochlorite (-OCl) to elucidate reliable the roles of -OCl in biological systems such as neutrophils. Peroxynitrite can be detected in distinction from nitric oxide and superoxide since APF does not react with nitric oxide, superoxide and hydrogen peroxide. Moreover, APF is resistant to light-induced autooxidation.

Synonym: Aminophenyl fluorescein, 2-[6-(4'-Amino)phenoxy-3H-xanthen-3-on-9-yl]benzoic acid.


Formula: C₂₆H₁₇NO₅
MW: 423.4 g/mol
Storage Temperature: Refrigerator
CAS Number: 359010-70-1
EINECS: 200-679-5
UN: 2265
ADR: 3,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/08/84/45390884.jpg
Order Now
Specification Test Results
Concentration ~5 mM
Formulation Dissolved in 0.47 mL dimethyl formamide
Purity ≥98% (HPLC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty