Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-P-Glycoprotein Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
801-002-C100
89153-160EA 1090 USD InStock
89153-160
Anti-P-Glycoprotein Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes an internal, highly conserved amino acid sequence (VQEALD and VQAALD) found in both protein isoforms, P-glycoprotein and MDR3 P-glycoprotein. Specific reactivity to human, mouse, rat, dog and primate. Detects bands of ~170kDa (MDR1 P-glycoprotein) and of ~140kDa (MDR3 P-glycoprotein) by Western blot.

Type: Primary
Antigen: P-GLYCOPROTEIN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty