Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MVP Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
801-005-C100 801-005-C050
89153-168EA 547.15 USD InStock
89153-168 89153-170
Anti-MVP Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2b

  • For Immunocytochemistry, Immunohistochemistry

Recognizes an internal epitope (P110) of human MVP/LRP, which is overexpressed in various human non-P-glycoprotein MDR tumor cell lines.

Type: Primary
Antigen: MVP
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty